Anti-FNTA Rabbit Polyclonal Antibody

Supplier: Abgent
AP2420A
80088-650EA 801.16 USD
80088-650
Anti-FNTA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FNTA (farnesyltransferase, CAAX box, alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR