Anti-MGAT1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2406A
80088-604EA 801.16 USD
80088-604
Anti-MGAT1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MGAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal ( 1–50 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR