Anti-MOGS Rabbit Polyclonal Antibody

Supplier: Abgent
AP2315A
80088-564EA 801.16 USD
80088-564
Anti-MOGS Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MOGS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal ( 1–50 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR