Anti-ITCH Rabbit Polyclonal Antibody

Supplier: Abgent
AP2171B
80088-472EA 763.01 USD
80088-472
Anti-ITCH Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ITCH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal ( 845–859 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR