Anti-SMURF1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2104A
80088-430EA 801.6 USD
80088-430
Anti-SMURF1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SMURF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal ( 1–100 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR