Anti-PJA Rabbit Polyclonal Antibody

Supplier: Abgent
AP2102C
80088-426EA 762.8 USD
80088-426
Anti-PJA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PJA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR