Anti-ACKR2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2012A
80088-298EA 801.16 USD
80088-298
Anti-ACKR2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ACKR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal ( 1–100 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR