Anti-BMP4 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1715A
80088-252EA 808.22 USD
80088-252
Anti-BMP4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BMP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal ( 1–50 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR