Anti-GJA8 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1548B
80088-118EA 801.16 USD
80088-118
Anti-GJA8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GJA8 (gap junction protein, alpha 8, 50kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR