Anti-GJB1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1541A
80088-092EA 763.01 USD
80088-092
Anti-GJB1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GJB1 (gap junction protein, beta 1, 32kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR