Anti-TLR6 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1506A
80088-030EA 801.6 USD
80088-030
Anti-TLR6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TLR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal ( 408–424 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR