Anti-APOBEC3G Rabbit Polyclonal Antibody

Supplier: Abgent
AP1351D
80087-996EA 801.16 USD
80087-996
Anti-APOBEC3G Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: APOBEC3G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR