Anti-RAD9 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1318A
80087-968EA 801.16 USD
80087-968
Anti-RAD9 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: 1–100 aa
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR