Anti-BCL2L Rabbit Polyclonal Antibody

Supplier: Abgent
AP1306A
80087-948EA 763.01 USD
80087-948
Anti-BCL2L Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-Bcl-x (BH3 Domain Specific) Reacts with Human

Type: Primary
Antigen: BCL2L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: 83–103 aa
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR