Anti-DNMT3L (DNA (cytosine-5-)-methyltransferase 3-like) Rabbit Polyclonal Antibody

Supplier: Abgent
AP1062A
80087-756EA 808.65 USD
80087-756
Anti-DNMT3L (DNA (cytosine-5-)-methyltransferase 3-like) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DNMT3L (DNA (cytosine-5-)-methyltransferase 3-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR