Anti-MBD3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1038A
80087-736EA 762.8 USD
80087-736
Anti-MBD3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MBD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: 100–200 aa
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR