Linear Pipette Stand for mLINE® Single Channel Mechanical Pipettors, Variable Volume, Sartorius
Supplier: Sartorius
725620
80087-478EA 268.31 USD
80087-478
Linear Pipette Stand for mLINE® Single Channel Mechanical Pipettors, Variable Volume, Sartorius
Pipette Stands
This pipette stand holds your pipettes in an upright position while not in use to avoid contamination from benches and tables.
Linear stand is designed for all Sartorius mechanical and electronic pipettes, particularly for mLINE®, Proline® Plus and Proline® mechanical devices. This stand is also compatible with a wide range of other pipette brands.