Sickle Cell Test Kits, ASI, Arlington Scientific
Supplier: ARLINGTON SCIENTIFIC INC.
The ASI Sickle Cell Test is a convenient and specific kit intended to be used as an aid in the qualitative detection of hemoglobin S (Hb-S) in anticoagulated whole blood. The test does not distinguish between sickle cell disease (HbS/S) and sickle cell trait (HbS/A). This test is not recommended for use on newborns under 3 months of age. Kit does not include controls.
- Accurate, easy and economical
- Detects both homozygous (S/S) and heterozygous (A/S) sickle cell
- Contains urea reagent for test confirmation
- Turbidity line test based upon modified Nalbandian procedure
- Room temperature storage
- Proudly made in USA
- Longest shelf life - up to 18 Months
- Sensitivity: 99%
- Specificity: 99%
Certifications: CE Marked
CPT Code 85660
510(k)-K960947
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...