VWR® Tissue Flotation Water Bath, 230 V
Supplier: VWR International
14792-2V
80086-984EA 1270.64 USD
80086-984
VWR® Tissue Flotation Water Bath, 230 V
Baths Tissue Baths
Typical use of tissue flotation water bath is in the preparation of tissue paraffin sections and the elimination of wrinkles and distortions of the tissue sections.
  • 230 V model, not for use in USA
  • Round and seamless
  • Sheet aluminum construction
  • Bath surface is black ceramic-coated steel
  • Improved visibility of tissue sections
Designed especially for tests and procedures using tissue cultures.
Round, seamless, sheet aluminum bath is insulated with fiberglass. It features a baked gray enamel exterior and a cover. Bath surface is black ceramic-coated steel, which offers improved visibility of tissue sections. The warm broad edge of the bath is a convenient drying surface for processed slides. Complete with thermostat reference dial and knob, thermometer holder, pilot light, 18-gauge three-wire rubber safety cord and plug, and skid-resistant feet.
Caution: 230 V model, not for use in USA. Corresponding model with voltage suitable for use in US (80086-982) is also available, please see separate product page.