Multi 3400i Handheld Multimeter, WTW
Supplier: WTW
2FA311
80086-946EA 7948.05 USD
80086-946
Multi 3400i Handheld Multimeter, WTW
Electrochemistry Meters pH and Multi-Parameter Meters
The WTW Multi-Parameter Field Meters allow you to read up to three parameters simultaneously.
  • Built-in memory for 500 data sets
  • Easy to use - automatic calibration
  • Large, bright LCD display
  • Energy saving automatic shut-off
  • Battery and sensor condition display
The completely waterproof instrument is extremely rugged and ideally suitable for field, laboratory or plant use.
CellOx® oxygen sensors are membrane-covered and waterproof. SenTix® pH electrodes feature optimized membrane glass that guarantees quick measurement, even at low temperatures. TetraCon® conductivity cells feature the highest degree of precision and linearity by optimized cell geometry.
Certifications: Waterproof to IP 66 & IP 67 and NEMA 4X and 6 UL, C-UL listed. CE marked.
Ordering information: Manufacturer's three-year warranty. Multimeters include a professional case with built-in measuring set-up, two beakers, two stands, carrying strap, line adapter power supply, calibration and maintenance supplies, and operating instructions. Additional sensors and accessories are also available; see 80085-412 series.