N-DEX® NightHawk® Gloves, Showa
Supplier: Showa
7700PFTS 7700PFTM 7700PFTL 7700PFTXL
80086-394CS 412.45 USD
80086-394 80086-396 80086-398 80086-400
N-DEX® NightHawk® Gloves, Showa
Gloves
These nonsterile, powder-free, textured finish ambidextrous gloves protect hands from a wide array of chemical hazards, while avoiding Type I and Type IV latex allergies.
  • 100% accelerator-free nitrile
  • Easy to put on and remove
  • Cuff prevents dirt from entering the glove
  • Surface provides tactile feel and optimal grip
  • Great freedom of movement for the wrist
  • Gloves feature ESD and static-dissipative properties
They are suitable for light assembly tasks, handling disposables, automotive applications, and chemical industries.
Certifications: EN 374; Cat. III; AQL 1,5.