GreenHouse Blowdown Evaporator, System 20, Heidolph Radleys
Supplier: Heidolph NA, LLC
15850200
80086-242EA 23911.6 USD
80086-242
GreenHouse Blowdown Evaporator, System 20, Heidolph Radleys
Evaporators Parallel Synthesis Systems
Turn your existing GreenHouse Parallel Synthesizer into a parallel evaporator, or simply use this system as your own personal benchtop evaporator
The GreenHouse Blowdown uses a precisely controlled flow of inert gas combined with digitally controlled heating to carefully evaporate your samples. Interchangeable plates with either 8 or 24 hollow pins deliver an equal flow of gas to each tube, vial or well. The absence of any vacuum avoids bumping, further protecting your sample during evaporation.
Ordering information: This System includes GreenHouse Blowdown head, Long Blowdown pin plate 6 x 4 for GreenHouse tubes, GreenHouse Blowdown base adaptor, Blowdown condenser, Blowdown 150 mL reservoir, Blowdown pin plate 6 x 4, Blowdown pin plate 4 x 2, Blowdown stand-alone base, Flowmeter 2 to 27 L/min with stand, Gas connection tubing 5 m, GreenHouse glass cylinder, AT01 vial rack 4 x 2 for 27.8 mm OD vials, AT02 vial rack 6 x 4 for 13 mm OD vials, AT03 vial rack 6 x 4 for 13.8 mm OD vials, AT05 vial rack 4 x 2 for 24.3 mm OD vials, AT06 vial rack 6 x 4 for 15 mm OD vials. Inert gas: Yes; Reflux: Yes.