GreenHouse Plus Parallel Synthesizer, System 27, Heidolph Radleys
Supplier: Heidolph NA, LLC
015850140
80086-224EA 22842.7 USD
80086-224
GreenHouse Plus Parallel Synthesizer, System 27, Heidolph Radleys
Evaporators Parallel Synthesis Systems
The patented GreenHouse Plus Parallel Synthesizer was designed with the help of scientists to perform up to 24 reactions in parallel with a volume range for 0.5 to 7 mL per reaction tube
Its removable reaction block holds 24 glass reaction tubes in the same position as a standard 24-well microplate, enabling the rapid transfer of samples to and from plates with a multichannel pipette or robotic pipetting system.
Ordering information: This system includes GreenHouse Plus Parallel Synthesizer with head, base, glass cylinder and two reaction blocks, GreenHouse pLuse reaction tube, 100pcs, GreeHouse Plus PTFE magnetic stirring bars, 40pcs, GreenHouse Plus sepra and sealing mat evaluation kit, 4pcs, Carousel Stirring Hotplate 115V, Carousel temperature controller.