pH Electrodes, SenTix® PLUS, WTW
Supplier: WTW
103648
80085-236EA 687.25 USD
80085-236 97042-812
pH Electrodes, SenTix® PLUS, WTW
Electrodes pH Electrodes
These proven and tested pH electrodes are easy to use and offer high quality and precision
SenTix® PLUS electrodes with gel electrolyte and a plastic body are robust and shock-proof. The membrane glass is optimally safe-guarded by a protective shield
SenTix® PLUS electrodes with liquid electrolyte – the capillary effect of the platinum wire guarantees constant outflow and prevents contamination by precipitates or dirt even during changes in temperature; with membrane glass for an even quicker response. SenTix® HW and HWS electrodes: Electrolyte flow speed can be adjusted for optimal use.
The optimised membrane glass guarantees quick measurement even at low temperatures. The easy to use fill-hole cover prevents electrolyte from leaking out during storage.