Anti-NME4 Rabbit Polyclonal Antibody

Supplier: Abgent
AP8153B
80085-062EA 800.94 USD
80085-062
Anti-NME4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NME4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (150–250 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR