Anti-GCKR Rabbit Polyclonal Antibody

Supplier: Abgent
AP8143B
80085-028EA 763.01 USD
80085-028
Anti-GCKR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GCKR (glucokinase (hexokinase 4) regulator)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR