Anti-PAPSS2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP8091B
80084-876EA 801.16 USD
80084-876
Anti-PAPSS2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PAPSS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR