Anti-PI3KC3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP8014B
80084-680EA 763.01 USD
80084-680
Anti-PI3KC3 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-PI 3 Kinase Class 3 (PI3KC3) (C-term) Reacts with Human

Type: Primary
Antigen: PI3KC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (871–887 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR