Anti-PI3KC3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP8014A
80084-678EA 801.6 USD
80084-678
Anti-PI3KC3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PI3KC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (39–54 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR