Anti-MLK4 alpha/beta Rabbit Polyclonal Antibody

Supplier: Abgent
AP8002A
80084-654EA 763.01 USD
80084-654
Anti-MLK4 alpha/beta Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-MLK4 alpha/beta (N-term) Reacts with Human

Type: Primary
Antigen: MLK4 alpha/beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (1–16 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR