Anti-MAP3K8 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7957B
80084-610EA 801.16 USD
80084-610
Anti-MAP3K8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAP3K8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (300–400 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR