Anti-SLK Rabbit Polyclonal Antibody

Supplier: Abgent
AP7951A
80084-600EA 763.01 USD
80084-600
Anti-SLK Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center (681–697 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR