Anti-PAK1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7926A
80084-550EA 763.01 USD
80084-550
Anti-PAK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (147–162 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR