Anti-MAP2K7 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7918A
80084-534EA 763.01 USD
80084-534
Anti-MAP2K7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAP2K7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (402–419 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR