Anti-MAP2K4 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7910A
80084-518EA 801.6 USD
80084-518
Anti-MAP2K4 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-MEKK4 (C-term) Reacts with Human

Type: Primary
Antigen: MAP2K4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (1081–1097 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR