Anti-MAP3K3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7909A
80084-516EA 763.01 USD
80084-516
Anti-MAP3K3 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-MEKK3 (N-term) Reacts with Human

Type: Primary
Antigen: MAP3K3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (124–141 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR