Anti-MAP2K5 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7906A
80084-510EA 763.01 USD
80084-510
Anti-MAP2K5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAP2K5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (425–438 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR