Anti-RAF1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7816A
80084-476EA 801.16 USD
80084-476
Anti-RAF1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (1–100 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR