Anti-TNK1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7722C
80084-412EA 808.65 USD
80084-412
Anti-TNK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TNK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center (200–300 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR