Anti-BRK (PTK6) Rabbit Polyclonal Antibody

Supplier: Abgent
AP7715A
80084-398EA 808.65 USD
80084-398
Anti-BRK (PTK6) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BRK (PTK6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (1–100 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR