Anti-ABL2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7695A
80084-360EA 763.01 USD
80084-360
Anti-ABL2 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-ABL2 (N-term) Reacts with Human

Type: Primary
Antigen: ABL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (1–100 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR