Anti-DOK5 (docking Protein 5) Rabbit Polyclonal Antibody

Supplier: Abgent
AP7693B
80084-356EA 801.16 USD
80084-356
Anti-DOK5 (docking Protein 5) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DOK5 (docking Protein 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (291–306 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR