Anti-NTRK1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7686A
80084-340EA 763.01 USD
80084-340
Anti-NTRK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NTRK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (772–787 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR