Anti-TEK (TIE2) Rabbit Polyclonal Antibody

Supplier: Abgent
AP7684A
80084-334EA 801.16 USD
80084-334
Anti-TEK (TIE2) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TEK (TIE2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (773–789 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR