Anti-DDR2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7683A
80084-332EA 763.01 USD
80084-332
Anti-DDR2 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-DDR2 (TYRO10) (C-term) Reacts with Human

Type: Primary
Antigen: DDR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (831–847 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR