Anti-RET Rabbit Polyclonal Antibody

Supplier: Abgent
AP7669B
80084-300EA 801.6 USD
80084-300
Anti-RET Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RET
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal T1078 (1078–1093 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR