Anti-PDGFRB Rabbit Polyclonal Antibody

Supplier: Abgent
AP7667C
80084-294EA 762.8 USD
80084-294
Anti-PDGFRB Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-PDGFRB (C-term P1028) Reacts with Human

Type: Primary
Antigen: PDGFRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal P1028
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR