Anti-EPHA4 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7609B
80084-182EA 801.6 USD
80084-182
Anti-EPHA4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EPHA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (890–904 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR