Anti-SRPK2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7553B
80084-154EA 801.16 USD
80084-154
Anti-SRPK2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SRPK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (400–500 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR