Anti-STK23 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7544C
80084-134EA 801.16 USD
80084-134
Anti-STK23 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: STK23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center (250–350 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR